Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin B, Free Acid, MP Biomedicals

Catalog No. ICN19516410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19516410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19516410 Supplier MP Biomedicals, Inc Supplier No. 0219516410

Chemical Identifiers

CAS 56360-46-4
Molecular Formula C20H6Br2N2O9-2
Molecular Weight (g/mol) 578.081
InChI Key ZBQZBWKNGDEDOA-UHFFFAOYSA-L
Synonym C.I. 45400
PubChem CID 7058116
ChEBI CHEBI:68617
IUPAC Name 4',5'-dibromo-2',7'-dinitro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)[N+](=O)[O-])[O-])Br)Br)[O-])[N+](=O)[O-]

Specifications

Assay Percent Range ∽ 85%
Formula Weight 580.1
Quantity 10 g
Chemical Name or Material Eosin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.