accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 56423-63-3

IUPAC Name: 2-bromopyrazine
Synonyms: 2-bromopyrazine
Molecular Weight (g/mol): 158.99
Molecular Formula: C4H3BrN2
InChi Key: WGFCNCNTGOFBBF-UHFFFAOYSA-N
SMILES: BrC1=NC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)