Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromopyrazine

Catalog No. CDS005022 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005022 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005022 Supplier Sigma Aldrich Supplier No. CDS005022100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-ethyl-2,4-pentanedione has been used in the preparation of:
  • N,n,n,n-tetradentate macrocyclic ligand
  • Vanadyl 3-ethylacetylacetonate, its effects in non-ketotic, streptozotocin-diabetic rats has been studied

Specifications

CAS 56423-63-3
Molecular Formula C4H3BrN2
Linear Formula C4H3BrN2
MDL Number MFCD08275680
Quantity 25 g
Synonym Bromopyrazine
Molecular Weight (g/mol) 158.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.