CAS RN 56425-91-3
CAS RN 56425-91-3
IUPAC Name:
2-methyl-1-(pyrimidin-5-yl)-1-[4-(trifluoromethoxy)phenyl]propan-1-ol
Synonyms:
flurprimidol
Molecular Weight (g/mol):
312.29
Molecular Formula:
C15H15F3N2O2
InChi Key:
VEVZCONIUDBCDC-UHFFFAOYNA-N
SMILES:
CC(C)C(O)(C1=CC=C(OC(F)(F)F)C=C1)C1=CN=CN=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Flurprimidol
CAS: 56425-91-3 Molecular Formula: C15H15F3N2O2