accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHCH3H3C

CAS RN 565-67-3

IUPAC Name: 2-methylpentan-3-ol
Synonyms: 2-methyl-3-pentanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: ISTJMQSHILQAEC-UHFFFAOYNA-N
SMILES: CCC(O)C(C)C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)