Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-pentanol, 97+%

Catalog No. AAL0801518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0801518 50 g
AAL0801509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0801518 Supplier Thermo Scientific Chemicals Supplier No. L0801518
  • $380.50
    $323.65 / Each of 1

    Save $56.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 565-67-3 | C6H14O | 102.177 g/mol

2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water

Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-67-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004567
InChI Key ISTJMQSHILQAEC-UHFFFAOYSA-N
Synonym 2-methyl-3-pentanol, 3-pentanol, 2-methyl, propanol, 1-isopropyl, acmc-20anqw, 4-methyl-3-pentanol, 2-methyl-pentan-3-ol, 4-01-00-01716 beilstein handbook reference, istjmqshilqaec-uhfffaoysa
PubChem CID 11264
IUPAC Name 2-methylpentan-3-ol
SMILES CCC(C(C)C)O

Specifications

Density 0.83
Boiling Point 127°C to 129°C
Flash Point 46°C (114°F)
Refractive Index 1.417
Quantity 50 g
UN Number UN2282
Beilstein 1718952
Solubility Information Amalgamate with ethanol,ethyl ether,benzene. Insoluble in water
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-pentanol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
EINECSNumber 209-286-3
RTECSNumber SA7400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.