accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5653-67-8

IUPAC Name: (2,3-dimethoxyphenyl)methanol
Synonyms: (2,3-dimethoxyphenyl)methanol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: CRLBBOBKCLYCJK-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CO)=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results