Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D176425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D176425G Supplier TCI America Supplier No. D176425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5653-67-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004612
InChI Key CRLBBOBKCLYCJK-UHFFFAOYSA-N
Synonym 2,3-dimethoxybenzyl alcohol, 2,3-dimethoxyphenyl methanol, o-veratryl alcohol, benzenemethanol, 2,3-dimethoxy, 2,3-dimethoxyphenyl methan-1-ol, 2,3-dimethoxybenzylalcohol, acmc-209lsx, ksc605g0n, benzenemethanol,2,3-dimethoxy, 2,3-dimethoxyphenyl methanol #
PubChem CID 79738
IUPAC Name (2,3-dimethoxyphenyl)methanol
SMILES COC1=CC=CC(=C1OC)CO

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 258°C
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethoxybenzyl Alcohol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.