accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56564-52-4

IUPAC Name: 2-(methylamino)-3-phenylpropanoic acid
Synonyms: phenylalaninenmethyl
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: SCIFESDRCALIIM-UHFFFAOYNA-N
SMILES: CNC(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)