accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH(E)

CAS RN 56578-18-8

IUPAC Name: (5E)-dec-5-en-1-ol
Synonyms: decan-5-en-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: WYPQHXVMNVEVEB-AATRIKPKSA-N
SMILES: CCCC\C=C\CCCCO
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)