Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-5-Decen-1-ol 95.0+%, TCI America™
SDP

Catalog No. D33085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33085G 5 g
D33081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33085G Supplier TCI America Supplier No. D33085G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56578-18-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00056187
InChI Key WYPQHXVMNVEVEB-AATRIKPKSA-N
Synonym trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038
PubChem CID 5283292
IUPAC Name (5E)-dec-5-en-1-ol
SMILES CCCC\C=C\CCCCO

Specifications

Color Colorless
Boiling Point 86°C
Quantity 5 g
Formula Weight 156.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-5-Decen-1-ol
EINECSNumber (2)-0258
RTECSNumber HE2096000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.