accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5661-71-2

IUPAC Name: azonane
Synonyms: 1H-azonine, octahydro-
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: NRHDCQLCSOWVTF-UHFFFAOYSA-N
SMILES: C1CCCCNCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results