Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octamethyleneimine 98.0+%, TCI America™
SDP

Catalog No. O02855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02855G Supplier TCI America Supplier No. O02855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5661-71-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00053378
InChI Key NRHDCQLCSOWVTF-UHFFFAOYSA-N
Synonym Azacyclononane, Octahydroazonine
PubChem CID 79742
IUPAC Name azonane
SMILES C1CCCCNCCC1

Specifications

Color Yellow
Boiling Point 188°C
Quantity 5 g
UN Number 1760
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Octamethyleneimine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.