accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(R)HN

CAS RN 56661-88-2

IUPAC Name: methyl (1R)-2-iminocyclohexane-1-carboxylate
Synonyms: methyl (1R)-2-iminocyclohexane-1-carboxylate
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: FDEZKFZZXQOWMM-ZCFIWIBFSA-N
SMILES: COC(=O)[C@@H]1CCCCC1=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results