Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle2-amino-1-cyclohexene-1-carboxylate, 99%, Thermo Scientific™

Catalog No. AC292760010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC292760010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292760010 Supplier Thermo Scientific Chemicals Supplier No. 292760010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56661-88-2
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H13NO2
Quantity 1g
MDL Number MFCD00216928
Synonym methyl 2-aminocyclohex-1-enecarboxylate, methyl 2-amino-1-cyclohexene-1-carboxylate, methyl 2-aminocyclohex-1-ene-1-carboxylate, maybridge3_000482, 2-amino-1-cyclohexene-1-carboxylic acid methyl ester, 1-cyclohexene-1-carboxylicacid, 2-amino-, methyl ester
InChI Key RMXRUKCQJHLQFA-UHFFFAOYSA-N
SMILES COC(=O)C1=C(CCCC1)N
IUPAC Name methyl 2-aminocyclohexene-1-carboxylate
Molecular Weight (g/mol) 155.197
PubChem CID 2724961
Formula Weight 155.2
Percent Purity ≥98.5%
Chemical Name or Material Methyl 2-amino-1-cyclohexene-1-carboxylate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.