accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CMgClCH3

CAS RN 5674-01-1

IUPAC Name: chloro(2-methylprop-2-en-1-yl)magnesium
Synonyms: chloro(2-methylprop-2-en-1-yl)magnesium
Molecular Weight (g/mol): 114.86
Molecular Formula: C4H7ClMg
InChi Key: BJVFGWYBOLMUEM-UHFFFAOYSA-M
SMILES: CC(=C)C[Mg]Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)