accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NCH3Br

CAS RN 56746-19-1

IUPAC Name: 4-bromo-2,6-diethylaniline
Synonyms: 4-bromo-2,6-diethylaniline
Molecular Weight (g/mol): 228.13
Molecular Formula: C10H14BrN
InChi Key: BEJYDMQQZUACPW-UHFFFAOYSA-N
SMILES: CCC1=CC(Br)=CC(CC)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results