Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-diethylaniline 98.0+%, TCI America™
SDP

Catalog No. B128410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B128410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B128410G Supplier TCI America Supplier No. B128410G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56746-19-1
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.133
MDL Number MFCD00059266
InChI Key BEJYDMQQZUACPW-UHFFFAOYSA-N
Synonym 2,6-Diethyl-4-bromoaniline
PubChem CID 611452
IUPAC Name 4-bromo-2,6-diethylaniline
SMILES CCC1=CC(=CC(=C1N)CC)Br

Specifications

Boiling Point 134°C
Quantity 10 g
Formula Weight 228.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2,6-diethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.