CAS RN 5687-48-9
IUPAC Name:
2-(2-acetamidoacetamido)acetic acid
Synonyms:
(2-acetamidoacetamido)acetic acid
Molecular Weight (g/mol):
174.16
Molecular Formula:
C6H10N2O4
InChi Key:
ZCASHLUDUSAKNN-UHFFFAOYSA-N
SMILES:
CC(=O)NCC(=O)NCC(O)=O
1
–
1
of
1
results
1
N-Acetylglycylglycine, 97%, Thermo Scientific™
CAS: 5687-48-9 Molecular Formula: C6H10N2O4 Molecular Weight (g/mol): 174.156 MDL Number: MFCD00037788 InChI Key: ZCASHLUDUSAKNN-UHFFFAOYSA-N Synonym: 2-2-acetamidoacetamido acetic acid,n-acetylglycylglycine,ac-gly-gly-oh,unii-01u1cr2e6l,2-acetamidoacetamido acetic acid,n-ac-gly-gly,dpgt,acetyl-gly-gly,acetyl-glycylglycine,glycine, n-n-acetylglycyl PubChem CID: 97591 IUPAC Name: 2-[(2-acetamidoacetyl)amino]acetic acid SMILES: CC(=O)NCC(=O)NCC(=O)O