accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56881-90-4

IUPAC Name: 3-hydroxyoxolan-2-one
Synonyms: 3-hydroxyoxolan-2-one
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: FWIBCWKHNZBDLS-UHFFFAOYNA-N
SMILES: OC1CCOC1=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)