Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-alpha-Hydroxy-gamma-butyrolactone 98.0+%, TCI America™
SDP

Catalog No. H09501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09501G 1 g
H0950100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09501G Supplier TCI America Supplier No. H09501G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56881-90-4
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00211246
InChI Key FWIBCWKHNZBDLS-UHFFFAOYNA-N
Synonym (R)-(+)-Dihydro-3-hydroxy-2(3H)-furanone, (R)-(+)-3-Hydroxytetrahydrofuran-2-one
PubChem CID 7157070
IUPAC Name 3-hydroxyoxolan-2-one
SMILES OC1CCOC1=O

Specifications

Color Yellow
Boiling Point 133°C
Quantity 1 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-alpha-Hydroxy-gamma-butyrolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.