accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOOO

CAS RN 5689-83-8

IUPAC Name: 4-methyl-1,3,2λ⁶-dioxathiolane-2,2-dione
Synonyms: 4-methyl-1,3,2λ⁶-dioxathiolane-2,2-dione
Molecular Weight (g/mol): 138.14
Molecular Formula: C3H6O4S
InChi Key: OQXNUCOGMMHHNA-UHFFFAOYNA-N
SMILES: CC1COS(=O)(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results