Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1,3,2-dioxathiolane 2,2-Dioxide, TCI America™
SDP

Catalog No. M247025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M247025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M247025G Supplier TCI America Supplier No. M247025G

missing translation for 'validMainframeMsg'

Specifications

CAS 5689-83-8
Color Brown-Yellow
Boiling Point 50°C
Molecular Formula C3H6O4S
MDL Number MFCD08703327
Quantity 25 g
Synonym 1,2-Propanediol Cyclic Sulfate, 1,2-Propylene Sulfate, 4-Methyl-2,2-dioxo-1,3,2-dioxathiolane
InChI Key OQXNUCOGMMHHNA-UHFFFAOYSA-N
SMILES CC1COS(=O)(=O)O1
IUPAC Name 4-methyl-1,3,2-dioxathiolane 2,2-dioxide
Molecular Weight (g/mol) 138.137
PubChem CID 9964107
Formula Weight 138.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1,3,2-dioxathiolane 2,2-Dioxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.