accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 569-41-5

IUPAC Name: 1,8-dimethylnaphthalene
Synonyms: 1,8-dimethylnaphthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: XAABPYINPXYOLM-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=CC=CC(C)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results