CAS RN 569-51-7
CAS RN 569-51-7
IUPAC Name:
benzene-1,2,3-tricarboxylic acid
Synonyms:
hemimellitic acid
Molecular Weight (g/mol):
210.14
Molecular Formula:
C9H6O6
InChi Key:
UJMDYLWCYJJYMO-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(C(O)=O)=C1C(O)=O
1
–
1
of
1
results
1
Hemimellitic Acid 98.0+%, TCI America™

CAS: 569-51-7 Molecular Formula: C9H6O6 Molecular Weight (g/mol): 210.141 MDL Number: MFCD00002468 InChI Key: UJMDYLWCYJJYMO-UHFFFAOYSA-N Synonym: 1,2,3-Benzenetricarboxylic Acid PubChem CID: 11288 IUPAC Name: benzene-1,2,3-tricarboxylic acid SMILES: C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O