Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemimellitic Acid 98.0+%, TCI America™
SDP

Catalog No. H15925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15925G 5 g
H159225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15925G Supplier TCI America Supplier No. H15925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 569-51-7
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.141
MDL Number MFCD00002468
InChI Key UJMDYLWCYJJYMO-UHFFFAOYSA-N
Synonym 1,2,3-Benzenetricarboxylic Acid
PubChem CID 11288
IUPAC Name benzene-1,2,3-tricarboxylic acid
SMILES C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 194°C
Color White
Quantity 5 g
Formula Weight 210.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hemimellitic Acid
EINECSNumber (3)-1279
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.