accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 569371-10-4

IUPAC Name: 4-{[3-(2-amino-2-carboxyethyl)-5-iodo-2,6-dioxo-1,2,3,6-tetrahydropyrimidin-1-yl]methyl}benzoic acid
Synonyms: 4-{[3-(2-amino-2-carboxyethyl)-5-iodo-2,6-dioxopyrimidin-1-yl]methyl}benzoic acid
Molecular Weight (g/mol): 459.2
Molecular Formula: C15H14IN3O6
InChi Key: JHSYCOCOIYSZGI-UHFFFAOYNA-N
SMILES: NC(CN1C=C(I)C(=O)N(CC2=CC=C(C=C2)C(O)=O)C1=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results