accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3C

CAS RN 5697-02-9

IUPAC Name: 2-methylnaphthalen-1-yl acetate
Synonyms: 2-methyl-1-naphthyl acetate
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: WVOAPRDRMLHUMI-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(C)C=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results