Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-naphthyl Acetate 98.0+%, TCI America™
SDP

Catalog No. M175725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M175725G 25 g
M1757250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M175725G Supplier TCI America Supplier No. M175725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5697-02-9
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
InChI Key WVOAPRDRMLHUMI-UHFFFAOYSA-N
Synonym Acetic Acid 2-Methyl-1-naphthyl Ester, 1-Acetoxy-2-methylnaphthalene
PubChem CID 11805727
IUPAC Name (2-methylnaphthalen-1-yl) acetate
SMILES CC1=C(C2=CC=CC=C2C=C1)OC(=O)C

Specifications

Melting Point 82°C
Quantity 25 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1-naphthyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.