CAS RN 56976-06-8
CAS RN 56976-06-8
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3-(tert-butylsulfanyl)propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3-(tert-butylsulfanyl)propanoic acid
Molecular Weight (g/mol):
277.38
Molecular Formula:
C12H23NO4S
InChi Key:
OGARKMDCQCLMCS-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-Cys But -OH, Advanced ChemTech
CAS: 56976-06-8 Molecular Formula: C12H23NO4S Molecular Weight (g/mol): 277.38 InChI Key: OGARKMDCQCLMCS-UHFFFAOYNA-N IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(tert-butylsulfanyl)propanoic acid SMILES: CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(O)=O