accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56990-57-9

IUPAC Name: (7E)-8-oxo-7-[2-(6-sulfonaphthalen-2-yl)hydrazin-1-ylidene]-7,8-dihydroquinoline-5-sulfonic acid
Synonyms: (7E)-8-oxo-7-[2-(6-sulfonaphthalen-2-yl)hydrazin-1-ylidene]quinoline-5-sulfonic acid
Molecular Weight (g/mol): 459.45
Molecular Formula: C19H13N3O7S2
InChi Key: BIQBDHSYQMHHPU-CJLVFECKSA-N
SMILES: OS(=O)(=O)C1=CC=C2C=C(N\N=C3/C=C(C4=CC=CN=C4C3=O)S(O)(=O)=O)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results