Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSC 87877, R&D Systems™
SDP

Catalog No. 261350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-135-0 Supplier R&D Systems Supplier No. 2613/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent inhibitor of shp2 and shp1 PTP

Potent inhibitor of shp2 and shp1 protein tyrosine phosphatases (PTP) (IC50 values are 0.318, 0.355, 1.691, 7.745, 65.617, 84.473 and 150.930 μM for shp2, shp1, PTP1B, HePTP, DEP1, CD45 and LAR respectively). Inhibits EGF-induced Erk1/2 activation in HEK293 cells and significantly reduces MDA-MB-468 cell viability/proliferation.

Chemical Identifiers

CAS 56990-57-9
Molecular Formula C19H13N3O7S2
Molecular Weight (g/mol) 459.447
InChI Key BIQBDHSYQMHHPU-CJLVFECKSA-N
Synonym 8-hydroxy-7-6-sulfo-naphthalen-2-ylazo-quinoline-5-sulfonic acid, 8-hydroxy-7-6-sulfo-2-naphthyl azo-5-quinolinesulfonic acid, ncistruc1_001562, ncistruc2_001738, 8-hydroxy-7-6-sulfonaphthalen-2-yl diazenyl-quinoline-5-sulfonic acid, 8-hydroxy-7-6-sulfonaphthalen-2-yl diazenyl quinoline-5-sulfonic acid, 8-hydroxy-7-e-6-sulfo-2-naphthyl azo quinoline-5-sulfonic acid, 7e-8-oxo-7-6-sulfonaphthalen-2-yl hydrazinylidene quinoline-5-sulfonic acid
PubChem CID 5459322
IUPAC Name (7E)-8-oxo-7-[(6-sulfonaphthalen-2-yl)hydrazinylidene]quinoline-5-sulfonic acid
SMILES C1=CC2=C(C(=O)C(=NNC3=CC4=C(C=C3)C=C(C=C4)S(=O)(=O)O)C=C2S(=O)(=O)O)N=C1

Specifications

Quantity 50 mg
Formula Weight 459.45
Percent Purity >98%
Product Line Tocris
Chemical Name or Material NSC 87877
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.