accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3H2N

CAS RN 57-14-7

IUPAC Name: 1,1-dimethylhydrazine
Synonyms: 1,1-dimethylhydrazine dimazine unsymmetrical dimethylhydrazine gem-dimethylhydrazine
Molecular Weight (g/mol): 60.1
Molecular Formula: C2H8N2
InChi Key: RHUYHJGZWVXEHW-UHFFFAOYSA-N
SMILES: CN(C)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results