accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOClClCl

CAS RN 57-15-8

IUPAC Name: 1,1,1-trichloro-2-methylpropan-2-ol
Synonyms: chlorobutanol chlorobutanol, anhydrous clorobutanol chlorobutanolum coliquifilm C4H7cl3O acetochlorone clorobutanolo chlorbutanolum khloreton
Molecular Weight (g/mol): 177.45
Molecular Formula: C4H7Cl3O
InChi Key: OSASVXMJTNOKOY-UHFFFAOYSA-N
SMILES: CC(C)(O)C(Cl)(Cl)Cl
Boiling Point 
  • (5)
Grade 
  • (5)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (4)
  • (5)
  • (5)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)