CAS RN 57-50-1
Thermo Scientific Chemicals Sucrose, ultrapure, 99%
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose (EP/BP/NF), Fisher Chemical™
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Thermo Scientific Chemicals Sucrose, 99%
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Thermo Scientific Chemicals Sucrose, Molecular Biology Grade
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Thermo Scientific Chemicals Sucrose, ACS
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose (Crystalline/Certified ACS), Fisher Chemical™
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
D-Sucrose (Molecular Biology), Fisher BioReagents
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Thermo Scientific Chemicals D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose, (Crystalline), MP Biomedicals
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N PubChem CID: 5988 ChEBI: CHEBI:17992 SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose, MP Biomedicals™
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: Beet sugar,Cane sugar,b-D-Fructofuranosyl-a-D-glucopyranoside,D-(+)-Saccharose IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose, 99.97%, MP Biomedicals™
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose, Ultrapure, 99.9%, Spectrum™ Chemical
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Sucrose, Crystal, ACS, Spectrum™ Chemical
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
D(+)-Sucrose, ACS reagent
CAS: 57-50-1 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00006626 InChI Key: CZMRCDWAGMRECN-PWPRYFECNA-N Synonym: sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand PubChem CID: 5988 ChEBI: CHEBI:17992 IUPAC Name: (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O