Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, MP Biomedicals™

Catalog No. MP04821721 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04821721 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP04821721 Supplier MP Biomedicals, Inc Supplier No. 04821721
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sucrose Ultra Pure, ≥99%, White Crystals

  • It is used as a source of carbon in media and animal diet.
  • 8.5% (w/v) sucrose was used in dielectrophoretic field-flow fractionation (dFFF) that allows the rapid and convenient analysis of bulk cell suspensions.
  • Sucrose is a non-reducing disaccharide composed of glucose and fructose linked via their anomeric carbons.
  • It is obtained commercially from sugarcane, sugar beet, and other plants and used extensively as a food and a sweetener.
  • The mixture of glucose and fructose is known as invert sugar.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym Beet sugar, Cane sugar, b-D-Fructofuranosyl-a-D-glucopyranoside, D-(+)-Saccharose
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 177°C to 187°C
Color White
Density 1.587 at 25°C (water = 1)
Quantity 5 kg
Beilstein 90825
Solubility Information Soluble in water (1g/0.5 mL at 25°C; 1g/0.2 mL boiling water), ethanol (1g/170 mL) or methanol (1g/100 mL); moderately soluble inglycerol or pyridine.
Optical Rotation 66.0°±5 (H2O, NH4OH)
Percent Purity ≥99%
Grade ≥99%
Physical Form Crystals
RTECSNumber WN6500000
storageNote1 Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.