accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5700-49-2

IUPAC Name: dihydrogen ethane-1,2-diamine diiodide
Synonyms: ethylenediamine dihydroiodide
Molecular Weight (g/mol): 315.93
Molecular Formula: C2H10I2N2
InChi Key: IWNWLPUNKAYUAW-UHFFFAOYSA-N
SMILES: [H+].[H+].[I-].[I-].NCCN
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)