Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethane-1,2-diammonium iodide

Catalog No. 9008525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-085-25G 5 g
90-085-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-085-25G Supplier Sigma Aldrich Supplier No. 9008525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Organohalide based perovskites have emerged as an important class of material for solar cell applications. Our perovskites precursors are useful for synthesizing mixed cation or anion perovskites needed for the optimization of the band gap, carrier diffusion length and power conversion efficiency of perovskites based solar cells.Product of Greatcell Solar(R).

Specifications

CAS 5700-49-2
Molecular Formula C2H8N2 -+ 2 HI
Linear Formula C2H8N2 -+ 2 HI
MDL Number MFCD00035535
Quantity 5 g
Synonym Ethylenediamine dihydriodide; 1,2-Ethanediamine dihydriodide; 1,2-Ethanediamine dihydroiodide; Ethane-1,2-diaminium iodide; Ethane-1,2-diamino dihydriodide; Ethylenediammonium diiodide; Greatcell Solar™.; 1,2-Diaminoethane dihydriodide
Molecular Weight (g/mol) 315.92
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.