accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57005-61-5

IUPAC Name: 5-methyl-6-oxo-1,6-dihydropyrazine-2,3-dicarbonitrile
Synonyms: 5-methyl-6-oxo-1H-pyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 160.14
Molecular Formula: C7H4N4O
InChi Key: GWQUFNJIVJLZRO-UHFFFAOYSA-N
SMILES: CC1=NC(C#N)=C(NC1=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results