CAS RN 5706-08-1
IUPAC Name:
4H-indeno[1,2-b]thiophen-4-one
Synonyms:
indeno[1,2-b]thiophen-4-one
Molecular Weight (g/mol):
186.23
Molecular Formula:
C11H6OS
InChi Key:
WSOJIQBCVMSHLB-UHFFFAOYSA-N
SMILES:
O=C1C2=C(SC=C2)C2=CC=CC=C12
1
–
1
of
1
results
1
4H-Indeno[1,2-b]thiophen-4-one 98.0+%, TCI America™
CAS: 5706-08-1 Molecular Formula: C11H6OS Molecular Weight (g/mol): 186.23 InChI Key: WSOJIQBCVMSHLB-UHFFFAOYSA-N PubChem CID: 12872762 IUPAC Name: indeno[1,2-b]thiophen-4-one SMILES: C1=CC=C2C(=C1)C3=C(C2=O)C=CS3