Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4H-Indeno[1,2-b]thiophen-4-one 98.0+%, TCI America™
SDP

Catalog No. I1003200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1003200MG 200 mg
I10031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1003200MG Supplier TCI America Supplier No. I1003200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5706-08-1
Molecular Formula C11H6OS
Molecular Weight (g/mol) 186.23
InChI Key WSOJIQBCVMSHLB-UHFFFAOYSA-N
PubChem CID 12872762
IUPAC Name indeno[1,2-b]thiophen-4-one
SMILES C1=CC=C2C(=C1)C3=C(C2=O)C=CS3

Specifications

Melting Point 101°C
Quantity 200 mg
Formula Weight 186.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4H-Indeno[1,2-b]thiophen-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.