accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH

CAS RN 5708-19-0

IUPAC Name: cyclohex-3-ene-1-carboxylic acid
Synonyms: cyclohex-3-ene-1-carboxylic acid
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: VUSWCWPCANWBFG-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC=CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results