Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Cyclohexene-1-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C32361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32361G Supplier TCI America Supplier No. C32361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5708-19-0
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD08275437
InChI Key VUSWCWPCANWBFG-ZCFIWIBFSA-N
Synonym (S)-(-)-1,2,3,6-Tetrahydrobenzoic Acid
PubChem CID 7128364
IUPAC Name (1S)-cyclohex-3-ene-1-carboxylic acid
SMILES C1CC(CC=C1)C(=O)O

Specifications

Melting Point 19°C
Color Colorless
Boiling Point 118°C
Quantity 1 g
UN Number 3265
Formula Weight 126.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-3-Cyclohexene-1-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.