accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5713-61-1

IUPAC Name: 2-(bromomagnesio)thiophene
Synonyms: 2-(bromomagnesio)thiophene
Molecular Weight (g/mol): 187.34
Molecular Formula: C4H3BrMgS
InChi Key: GSHPYJFNTAMRJF-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=CS1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results