Learn More
2-Thienylmagnesium bromide, 1M solution in THF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 65.0°C |
| Color | Brown to Yellow |
| CAS Min % | 17.6, 79.62 |
| Physical Form | Solution |
| CAS Max % | 20.38, 82.4 |
| Quantity | 50 mL |
| Flash Point | −21°C |
| Packaging | AcroSeal™ Glass Bottle |
| Specific Gravity | 1.011 |
| Density | 1.0110g/mL |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides. May cause respiratory irritation. Suspected of causing cancer.<br/ |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.