accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOHOHONaNa(E/Z)

CAS RN 5715-76-4

IUPAC Name: 4-[(3-carboxy-4-oxo-1,4-dihydronaphthalen-1-ylidene)(phenyl)methyl]-1-hydroxynaphthalene-2-carboxylic acid disodium
Synonyms: 4-[(3-carboxy-4-oxonaphthalen-1-ylidene)(phenyl)methyl]-1-hydroxynaphthalene-2-carboxylic acid disodium
Molecular Weight (g/mol): 508.44
Molecular Formula: C29H18Na2O6
InChi Key: DAUJJPVGSGIGFT-UHFFFAOYSA-N
SMILES: [Na].[Na].OC(=O)C1=CC(C(C2=CC=CC=C2)=C2C=C(C(O)=O)C(=O)C3=CC=CC=C23)=C2C=CC=CC2=C1O
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)