Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthochrome Green, Thermo Scientific™

Catalog No. AC195740250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC195740250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC195740250 Supplier Thermo Scientific Chemicals Supplier No. 195740250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5715-76-4
Molecular Formula C29H18Na2O6
Molecular Weight (g/mol) 508.437
InChI Key DAUJJPVGSGIGFT-UHFFFAOYSA-N
Synonym C.I. 44530
PubChem CID 131881316
IUPAC Name 4-[(3-carboxy-4-oxonaphthalen-1-ylidene)-phenylmethyl]-1-hydroxynaphthalene-2-carboxylic acid;sodium
SMILES C1=CC=C(C=C1)C(=C2C=C(C(=O)C3=CC=CC=C32)C(=O)O)C4=CC(=C(C5=CC=CC=C54)O)C(=O)O.[Na].[Na]

Specifications

Beilstein 10, II, 762
Infrared Spectrum Authentic
Formula Weight 506.41
UV Spectrometry Complies
Color Brown to Brown/Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Naphthochrome Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.