accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57212-58-5

IUPAC Name: potassium 2-({[5-(hydroxymethyl)-2-methyl-3-oxo-3,4-dihydropyridin-4-ylidene]methyl}amino)-3-methylpentanoate
Synonyms: potassium 2-({[5-(hydroxymethyl)-2-methyl-3-oxopyridin-4-ylidene]methyl}amino)-3-methylpentanoate
Molecular Weight (g/mol): 318.41
Molecular Formula: C14H19KN2O4
InChi Key: DCFBAHPGYZGVEW-UHFFFAOYNA-M
SMILES: [K+].CCC(C)C(NC=C1C(CO)=CN=C(C)C1=O)C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results