Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxylidene-L-isoleucine Potassium Salt, TCI America™
SDP

Catalog No. P09891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09891G Supplier TCI America Supplier No. P09891G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57212-58-5
Molecular Formula C14H19KN2O4
Molecular Weight (g/mol) 318.414
MDL Number MFCD00059937
InChI Key DCFBAHPGYZGVEW-MOLPEXLISA-M
PubChem CID 91873236
IUPAC Name potassium;(2S,3S)-2-[[(E)-[5-(hydroxymethyl)-2-methyl-3-oxopyridin-4-ylidene]methyl]amino]-3-methylpentanoate
SMILES CCC(C)C(C(=O)[O-])NC=C1C(=CN=C(C1=O)C)CO.[K+]

Specifications

Quantity 1 g
Formula Weight 318.41
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxylidene-L-isoleucine Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.