accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)OO(S)HNH3COHOOCH3

CAS RN 57224-63-2

IUPAC Name: methyl (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms: methyl (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate|Z-THR-OME
Molecular Weight (g/mol): 267.28
Molecular Formula: C13H17NO5
InChi Key: OPZWAOJFQFYYIX-KOLCDFICSA-N
SMILES: COC(=O)[C@@H](NC(=O)OCC1=CC=CC=C1)[C@@H](C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)